Benzylalkohol ≥99,5%, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

9050-01EA 151 CHF
9050-01
Benzylalkohol ≥99,5%, BAKER ANALYZED®, J.T.Baker®
Benzylalkohol
Formel: C₆H₅CH₂OH
Siedepunkt: 205 °C (1013 hPa)
Melting Pt: –15.3 °C
Dichte: 1.05 g/cm³ (20 °C)
Flash-Pt: 101 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004599
CAS-Nummer: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use = 99.5 %
Assay (C₆H₅CH₂OH) (by GC, corrected for water) ≥99.5 %
Color (APHA) ≤10
Residue after Ignition ≤0.005 %
Solubility in H₂O Passes Test
Chlorinated Compounds (as Cl) Passes Test
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR