5-Carboxy-X-rhodamine succinimidyl ester (5-ROX SE) dye_Amine-reactive fluorescent

Lieferant: Biotium

Synonyms: 5-Carboxy-X-rhodamine, NHS ester, 5-ROX, SE, Carboxy-X-rhodamine succinimidyl ester, 5 isomer

90036
90036.EA 390 CHF
90036.
5-Carboxy-X-rhodamine succinimidyl ester (5-ROX SE) dye_Amine-reactive fluorescent
5-Carboxy-X-rhodamine succinimidyl ester (5-ROX SE)

5-ROX, SE (5-Carboxy-X-rhodamine, succinimidyl ester, single isomer) is the amine-reactive form of 5-carboxy-X-rhodamine single isomer.


  • λEx/λ Em(MeOH) = 568/595 nm
  • Dark red solid soluble in DMSO or DMF
  • Store at -20°C and protect from light

Formel: C₃₇H₃₃N₃O₇
MW: 631.67 g/mol
Lager Temperatur: Gefrierschrank
CAS-Nummer: 344402-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7d44cb616f929f31a6d86b178c88520b038de55&vid=c7d44cb616f929f31a6d86b178c88520b038de55&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR