Natriumborhydrid 4.4 M aqueous solution stable

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

87659.AE 87659.AP 87659.K3
87659.AEEA 60.5 CHF
87659.AE 87659.AP 87659.K3
Natriumborhydrid 4.4 M aqueous solution stable
Natriumtetrahydroborat

CAS No.: 16940-66-2 Molecular Formula: BH4Na Formula Weight: 37.83 Physical Form: Liquid Density: 1.4 Sensitivity: Air Sensitive Appearance: Colourless UN No.: UN3320 MDL No.: MFCD00003518

Formel: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400
Dichte: 1.4
Flash-Pt: 69
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003518
CAS-Nummer: 16940-66-2
EINECS: 241-004-4
UN: 3320
ADR: 8 II
Merck Index: 13,08664

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR