Tetrabutylammoniumacetat, Supelco®

Lieferant: Merck

Synonyms: Tetra-n-butylammonium acetate

MQ100

86835-25G
86835-25GEA 475 CHF
86835-25G
Tetrabutylammoniumacetat, Supelco®
Tetrabutylammoniumacetat
Formel: C₁₈H₃₉NO₂
MW: 301.51 g/mol
MDL Number: MFCD00043208
CAS-Nummer: 10534-59-5
EINECS: 234-101-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5704e67222ad80e05ebdc855205be36b515b5a14&vid=e7399998b4e1019c403127696133c0ce67f47d2d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR