Rosolsäure (Aurin) 84% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyms: 4-(Bis(4-hydroxyphenyl)methylene)cyclohexa-2,5-dien-1-one

M-Clarity™ quality level = MQ200

861324-100G 861324-25G
861324-100GEA 368 CHF
861324-100G 861324-25G
Rosolsäure (Aurin) 84% (Farbstoffgehalt), Sigma-Aldrich®
Rosolsäure (Aurin)

p-Rosolic acid (C.I. 43800) is an acid-baseindicator from the triphenylmethane group. It is also known as aurin and 4-[Bis(4-hydroxyphenyl)methylene]-2,5-cyclohexadienone.


  • p-Rosolic acid has been used to induce cellular stress in HEK293 cells
  • The transition range of p-Rosolic acid is around pH 6,6 to 8,0 (Below this pH range it is yellow and above it red)

Formel: C₁₉H₁₄O₃
Lager Temperatur: Room Temperature
MDL Number: MFCD00001624
CAS-Nummer: 603-45-2
EINECS: 210-041-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48fbe4713b9e095325724d327711b1cb0b894df6&vid=fceb30068c47049a22436f96f4f215cde9e3c88e&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength 264 - 270 nm
Extinction Coefficient ≥50000
Extinction Coefficient, Acidified Ethanol ≥11000
Carbon Content 66.0 - 90.8 %
Visual Interval, pH 6.6 Yellow-Orange Pass
Visual Interval, pH 8.0 Pink to Red Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR