tri-Natriumphosphat Dodecahydrat 98,0-102,0%, AnalaR NORMAPUR® Reag. Ph.Eur., ACS

Lieferant: VWR Chemicals
Achtung

Synonyms: Sodium Phosphate tribasic 12-Hydrate, Sodium phosphate dodecahydrate, Sodium phosphate tribasic dodecahydrate

85742.290EA 105 CHF
85742.290 85742.360 85742.460
tri-Natriumphosphat Dodecahydrat 98,0-102,0%, AnalaR NORMAPUR® Reag. Ph.Eur., ACS
tri-Natriumphosphat Dodecahydrat
Formel: Na₃PO₄·12H₂O
MW: 380.12 g/mol
Melting Pt: ∼75 °C
Dichte: 1.62 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149198
CAS-Nummer: 10101-89-0
EINECS: 231-509-8
Merck Index: 13,08736

Order Now

Spezifikation Testergebnisse

Assay                            98.0 - 102.0 %
Excess alkali (as NaOH) Max. 2.5 %
Heavy metals (as Pb) Max. 10 ppm
Insoluble matter Max. 100 ppm
Chloride (Cl) Max. 10 ppm
Sulphate (SO₄) Max. 100 ppm
Iron (Fe)    Max. 10 ppm
Conforms to Reag. Ph.Eur./ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR