Ethylacetat ≥99,9%, HiPerSolv CHROMANORM® für die LC-MS

Lieferant: VWR Chemicals
Gefahr

Synonyms: Acetic acid ethyl ester

Filtriert über 0,2 µm-Filter. Unter Stickstoff abgefüllt.

85481.320EA 90.6 CHF
85481.320
Ethylacetat ≥99,9%, HiPerSolv CHROMANORM® für die LC-MS
Ethylacetat
Formel: CH₃COOC₂H₅
MW: 88.11 g/mol
Siedepunkt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Dichte: 0.902 g/cm³ (20 °C)
Flash-Pt: –4 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009171
CAS-Nummer: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance Clear colorless liquid
Acidity (as Acetic acid) max. 0.003 %
Alkalinity (as Ammonia) max. 0.0005 %
Color (APHA, Pt-Co) max. 10
Assay (GC, on anhydrous basis) min. 99.9 %
Residue after evaporation max. 0.0005 %
Water (KF) max. 0.05 %
T260nm min. 75 %
T275nm min. 98 %
LC-MS suitability test Complies
Ag (Silver) max. 0.1 ppm
Al (Aluminum) max. 0.05 ppm
Ba (Barium) max. 0.1 ppm
Bi (Bismuth) max. 0.1 ppm
Ca (Calcium) max. 0.05 ppm
Cd (Cadmium) max. 0.05 ppm
Co (Cobalt) max. 0.05 ppm
Cr (Chromium) max. 0.02 ppm
Cu (Copper) max. 0.02 ppm
Fe (Iron) max. 0.02 ppm
K (Potassium) max. 0.05 ppm
Li (Lithium) max. 0.1 ppm
Mg (Magnesium) max. 0.05 ppm
Mn (Manganese) max. 0.02 ppm
Mo (Molybdenum) max. 0.05 ppm
Na (Sodium) max. 0.05 ppm
Ni (Nickel) max. 0.02 ppm
Pb (Lead) max. 0.02 ppm
Sn (Tin) max. 0.05 ppm
Sr (Strontium) max. 0.05 ppm
Zn (Zinc) max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR