2-Propanol 99,9, PESTINORM® Supra Trace Trace_analysis_organic

Lieferant: VWR Chemicals
Gefahr

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

Filter 0,2 um_Inert_Gas_filled

85391.320EA 226 CHF
85391.320
2-Propanol 99,9, PESTINORM® Supra Trace Trace_analysis_organic
2-Propanol
Formel: (CH₃)₂CHOH
MW: 60.1 g/mol
Siedepunkt: 82
Melting Pt: –89 °C
Dichte: 0.786 g/cm³ (20 °C)
Flash-Pt: 12 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011674
CAS-Nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Assay (on anhydrous substance) Min. 99.9 %
Evaporation residue Max 0.0003%
Colouration Max 10 APHA
Water Max 0.05%
GC/ECD Dioxins, Furans & PCB's Max 5 ng/l
GC/ECD any pesticide (as Lindane) Max 5 ng/l
GC/NPD any pesticide (as Parathion) Max 10 ng/l
GC/ECD 1,2,4-TCB to daca-PCB (as Lindane) Max 5 pg/ml
GC/ECD DCM to 1,2,4-TCB (as TCM) Max 2 ng/l
GC/FID C10 to C40 (as n-Decane) Max 2 ng/l
GC/MSD C10 to C40 (as Decane; 30-600amu) Max 2 ng/l
Filtered through 0.2um, filled under inert gas Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR