, HiPerSolv CHROMANORM® LC-MS

Lieferant: VWR Chemicals
Gefahr


Nitrogen_filled

84866.290EA 144 CHF
84866.290 84866.320
, HiPerSolv CHROMANORM® LC-MS
Acetonitril mit 0,1% (v/v) Ameisensäure
Lager Temperatur: Raumtemperatur
EINECS: 200-835-2
UN: 1648
ADR: 3 II

Order Now

Spezifikation Testergebnisse

Nominal concentration (V/V) 0.095 - 0.105 %
Evaporation residue Max. 0.0001 %
Water Max. 0.005 %
Ca (Calcium) Max. 0.2 ppm
K (Potassium) Max. 0.2 ppm
Mg (Magnesium) Max. 0.2 ppm
Na (Sodium) Max. 0.5 ppm
Transmittance (235 nm) Min. 20 %
Transmittance (245 nm) Min. 50 %
Transmittance (250 nm) Min. 80 %
Transmittance (260 nm) Min. 95 %
Transmittance (270 nm) Min. 99 %
Suitable for LC-MS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR