Titan(IV)oxid 99,0-100,5, AnalaR NORMAPUR®

Lieferant: VWR Chemicals

Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide

84853.290EA 121 CHF
84853.290 84853.460
Titan(IV)oxid 99,0-100,5, AnalaR NORMAPUR®
Titan(IV)oxid
Formel: TiO₂
MW: 79.88 g/mol
Siedepunkt: 2900
Melting Pt: 1855 °C
Dichte: 4.2 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011269
CAS-Nummer: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549

Order Now

Spezifikation Testergebnisse

Assay 99.0 - 100.5 %
Heavy metals (as Pb) Max. 20 ppm
Loss on drying (105°C) Max. 0.5 %
Loss on ignition (800°C) Max. 0.5 %
Solubility in hot water Max. 0.5 %
Solubility in hydrochloric acid Max. 0.5 %
As (Arsenic) Max. 1 ppm
Fe (Iron) Max. 0.02 %
Sb (Antimony) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR