Methanol, BAKER ANALYZED®, isocratic HPLC, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

8404.1000EA 45.8 CHF
8404.1000 8404.2500 8404.5000 8404.9030RC
Methanol, BAKER ANALYZED®, isocratic HPLC, J.T.Baker®
Methanol
Formel: CH₃OH
MW: 32.04 g/mol
Siedepunkt: 64.6
Melting Pt: –98 °C
Dichte: 0.7918 g/cm³ (20 °C)
Flash-Pt: 11 °C (geschlossener Tiegel)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004595
CAS-Nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Assay min. 99.8 %
Color (APHA) max. 10
Residue after Evaporation (in ppm) max. 5
Titrable Acid (meq/g) max. 0.0003
Titrable Base (meq/g) max. 0.0002
Water (H2O) max. 0.05 %
Ultraviolet Absorbance (1.00-cm path vs water):
at 225 nm max. 0.30
at 240 nm max. 0.10
at 265 nm max. 0.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR