Ferrozine mono-sodium salt hydrate, Supelco®

Lieferant: Merck

Synonyms: FerroZine iron reagent hydrate, 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulphonic acid monosodium salt hydrate, 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4',4''-disulphonic acid monosodium salt hydrate, PDT disulphonate monosodium salt hydrate

M-Clarity™ Qualitätsstufe = MQ100

82950-1G 82950-5G
82950-1GEA 80.7 CHF
82950-1G 82950-5G
Ferrozine mono-sodium salt hydrate, Supelco®
Ferrozine mono-sodium salt hydrate
Formel: C₂₀H₁₃N₄NaO₆S₂·H₂O
MW: 492.47 g/mol (anhy)
MDL Number: MFCD00078417
CAS-Nummer: 63451-29-6
EINECS: 274-196-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=827c458da2852cd8c20c3ba00af19dbb3823ba8c&vid=827c458da2852cd8c20c3ba00af19dbb3823ba8c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR