Diethylether Stabilised, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

8254.1000EA 87.3 CHF
8254.1000 8254.1000S 8254.5000 8254.9025
Diethylether Stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Diethylether
Formel: (CH₃CH₂)₂O
MW: 74.12 g/mol
Siedepunkt: 35
Melting Pt: –123 °C
Dichte: 0.7135 g/cm³ (20 °C)
Flash-Pt: –40 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011646
CAS-Nummer: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Exceeds ACS Specifications
Assay (by GC) min. 99.5 %
Alcohol (C2H5OH) max. 0.01 %
Carbonyl Compounds (as HCHO) max. 0.001 %
Color (APHA) max. 10
Density (g/ml) at 20°C 0.712 - 0.714
Foreign Odor passes test
Peroxide (as H2O2) max. 3 ppm
Residue after Evaporation max. 0.001 %
Substances Darkened by H2SO4 passes test
Titrable Acid (meq/g) max. 0.0002
Water (H2O) max. 0.01 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR