Petroleumbenzin, 80…110 °C, BAKER, Laboratory Reagent, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

8242.1000EA 54.1 CHF
8242.1000
Petroleumbenzin, 80…110 °C, BAKER, Laboratory Reagent, J.T.Baker®
Petroleumbenzin
Siedepunkt: 80…110 °C (1013 hPa)
Dichte: 0.7
Flash-Pt: -10
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00081849
CAS-Nummer: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II

Order Now

Spezifikation Testergebnisse

Boiling Range 80 - 110 °C
Residue after Evaporation max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR