Reinraum-Klebebänder mit leitfähigem Raster, CGT™
Lieferant: MICRONOVA
CGT-12 CGT-34_U CGT-2 CGT-6 CGT-1 CGT-15 CGT-38
819-0074EA 32.7 CHF
819-0074 819-0077 819-0076 819-0079 819-0018 819-0075 819-0078
Reinraum-Klebebänder mit leitfähigem Raster, CGT™
Klebebänder
Conductive, anti-static polypropylene tape, specially designed with conductive grids embedded between two anti-static layers, for use on devices in ESD sensitive environments.
  • Heavy-duty, anti-static copolymer tape with acrylic adhesive
  • Manufactured for use in ESD sensitive environments
  • Features a conductive grid embedded between two antistatic layers
  • Conducts charge away from substrate
  • Protects sensitive devices that would be otherwise damaged by normal insulating tape
  • Provides an effective static shield and prevents static buildup
  • Prevents shedding,
More Product Information