Lucifer yellow CH dilithium salt fluorescent dye, Histology

Lieferant: Biotium

Synonyms: 6-Amino-2,3-dihydro-1,3-dioxo-2-hydrazinocarbonylamino-1H-benz[d,e]isoquinoline-5,8-disulphonic acid dilithium salt, LY CH lithium salt

80015
80015.EA 167 CHF
80015.
Lucifer yellow CH dilithium salt fluorescent dye, Histology
Lucifer yellow CH dilithium salt

Lucifer Yellow CH lithium salt (LY CH lithium salt) is a widely used polar molecular tracer for studying neuronal morphology. The fluorescent molecule contains a carbohydrazide that allow the molecule to be aldehyde-fixable.


  • λEx/λ Em = 428/536 nm
  • Yellow solid soluble in water
  • Store at 4°C and protect from light

Formel: C₁₃H₉N₅O₉S₂Li₂
MW: 457.25 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD00006922
CAS-Nummer: 67769-47-5
EINECS: 267-047-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a37d0e54e33266ea24800068e5ef261e5fd81e0&vid=4a37d0e54e33266ea24800068e5ef261e5fd81e0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR