tert-Butylethylether zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyms: 2-Ethoxy-2-methylpropane

M-Clarity™ quality level = MQ200

8.41873.0005 8.41873.0025 8.41873.0002
8.41873.0005EA 49.7 CHF
8.41873.0005 8.41873.0025 8.41873.0002
tert-Butylethylether zur Synthese, Sigma-Aldrich®
tert-Butylethylether
Formel: C₆H₁₄O
Siedepunkt: 71 °C (1013 hPa)
Melting Pt: –97 °C
Dichte: 0.752 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009225
CAS-Nummer: 637-92-3
EINECS: 211-309-7
UN: 1179
ADR: 3,II
Merck Index: 13,03809

Order Now

Spezifikation Testergebnisse

Solubility 12 g/l (25 °C)
Melting Point -97 °C
Molar Mass 102.18 g/mol
Boiling Point 71 °C (1013 hPa)
Vapor Pressure 173 hPa (25 °C)
Flash Point -19 °C
Explosion Limit 1.23 - 7.7 %(V)
Density 0.74 g/cm3 (20 °C)
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR