Phosphor(V)chlorid zS, Sigma-Aldrich®

Lieferant: Merck
Gefahr


MQ200

8.22340.0100 8.22340.0500 8.22340.1000 8.22340.9025
8.22340.0100EA 55.7 CHF
8.22340.0100 8.22340.0500 8.22340.1000 8.22340.9025
Phosphor(V)chlorid zS, Sigma-Aldrich®
Phosphor(V)chlorid
Formel: Cl₅P
MW: 208.24 g/mol
Melting Pt: ∼148 °C
Dichte: 3.6 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011439
CAS-Nummer: 10026-13-8
EINECS: 233-060-3
UN: 1806
ADR: 8 II
Merck Index: 12,07508

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) soluble,(decomposition)
Melting Point 148 °C
Molar Mass 208.24 g/mol
Boiling Point 167 °C (decomposition)
Vapor Pressure 0.016 hPa (20 °C)
Density 2.1 g/cm3 (20 °C)
pH (H2O) acidic
Hazard Class 8
Hazard Class Description Corrosive material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR