1,8-Dihydroxy-9,10-anthrachinon zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyms: Dantron, Danthron, 1,8-Dihydroxyanthraquinone

M-Clarity™ quality level = MQ200

8.22028.9025
8.22028.9025EA 0 CHF
8.22028.9025
1,8-Dihydroxy-9,10-anthrachinon zur Synthese, Sigma-Aldrich®
1,8-Dihydroxy-9,10-anthrachinon
Formel: C₁₄H₈O₄
Melting Pt: 190…193 °C
Dichte: 1.537 g/cm³ (20 °C)
Flash-Pt: >200 °C
MDL Number: MFCD00001211
CAS-Nummer: 117-10-2
EINECS: 204-173-5
Merck Index: 13,02841

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) insoluble
Melting Point 190 - 193 °C
Molar Mass 240.21 g/mol
Bulk Density 600 kg/m3
Flash Point 200 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR