1-Naphthylamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyms: 1-Naphthylammonium chloride

M-Clarity™ quality level = MQ200

8.20866.0100
8.20866.0100EA 121 CHF
8.20866.0100
1-Naphthylamin Hydrochlorid, Sigma-Aldrich®
1-Naphthylamin Hydrochlorid
Formel: C₁₀H₁₀ClN
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00036370
CAS-Nummer: 552-46-5
EINECS: 209-014-3
UN: 2811
ADR: 6.1,II

Order Now

Spezifikation Testergebnisse

Solubility 37.7 g/l (20 °C)
Melting Point 272 - 275 °C
Molar Mass 179.65 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR