Dinatriumfumarat zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyms: Disodium Fumarate, Fumaric acid disodium salt, Fumaric Acid Disodium Salt, Sodium fumarate

M-Clarity™ quality level = MQ200

8.20584.0100 8.20584.0500 8.20584.1000 8.20584.9025
8.20584.0100EA 31.4 CHF
8.20584.0100 8.20584.0500 8.20584.1000 8.20584.9025
Dinatriumfumarat zur Synthese, Sigma-Aldrich®
Dinatriumfumarat
Formel: C₄H₂Na₂O₄
MDL Number: MFCD00064567
CAS-Nummer: 17013-01-3
EINECS: 241-087-7

Order Now

Spezifikation Testergebnisse

Solubility 228 g/l (25 °C)
Molar Mass 160.04 g/mol
pH 7 - 8 (50 g/l, H2O)
Synonyms Fumaric acid disodium salt
Hill Formula C4H2Na2O4
Chemical Formula NaOOCCH=CHCOONa


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR