tert-Butylmethylether zur Synthese, Supelco®

Lieferant: Merck
Gefahr

Synonyms: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether

8.18109.9025
8.18109.9025EA 0 CHF
8.18109.9025
tert-Butylmethylether zur Synthese, Supelco®
tert-Butylmethylether
Formel: (CH₃)₃COCH₃
MW: 88.15 g/mol
Siedepunkt: 55.2
Melting Pt: –108.6 °C
Dichte: 0.7405 g/cm³ (20 °C)
Flash-Pt: –28 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008812
CAS-Nummer: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3 II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR