Hydrogen fluoride pyridine complex (HF 65%) zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyms: Olah′s reagent, Pyridinium polybifluoride, Pyridine hydrofluoride, HF-Pyridine, PPHF, Poly(pyridine fluoride), Pyridinium poly(hydrogen fluoride)

M-Clarity™ Qualitätsstufe = MQ200

8.18068.0100
8.18068.0100EA 239 CHF
8.18068.0100
Hydrogen fluoride pyridine complex (HF 65%) zur Synthese, Sigma-Aldrich®
Hydrogen fluoride pyridine complex
Formel: C₅H₅N·(HF)x
Lager Temperatur: Kühlschrank
MDL Number: MFCD00012436
CAS-Nummer: 62778-11-4
EINECS: 231-634-8
UN: 1790
ADR: 8 (6.1) I

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Density 1.14 g/cm3 (20 °C)
pH  - 7 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR