Cetrimoniumbromid zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

M-Clarity™ quality level = MQ200

8.14119.0100 8.14119.0500
8.14119.0100EA 39.3 CHF
8.14119.0100 8.14119.0500
Cetrimoniumbromid zur Synthese, Sigma-Aldrich®
Cetrimoniumbromid
Formel: CH₃(CH₂)₁₅N(Br)(CH₃)₃
Melting Pt: 237…243 °C
Flash-Pt: 244 °C (closed cup)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011772
CAS-Nummer: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9,III
Merck Index: 13,02034

Order Now

Spezifikation Testergebnisse

Solubility 0.192 g/l (20 °C)
Melting Point 250 - 256 °C
Molar Mass 364.45 g/mol
Bulk Density 390 kg/m3
pH 5 - 7 (50 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR