1,1,1-Tris(hydroxymethyl)propan, Sigma-Aldrich®

Lieferant: Merck

Synonyms: 1,1,1-Trimethylolpropane, Propylidynetrimethanol, Trimethylol propane, Propylidynetris(methanol), 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol

M-Clarity™ quality level = MQ200

8.08394.0100 8.08394.1000 8.08394.2500 8.08394.9050
8.08394.0100EA 68.4 CHF
8.08394.0100 8.08394.1000 8.08394.2500 8.08394.9050
1,1,1-Tris(hydroxymethyl)propan, Sigma-Aldrich®
1,1,1-Tris(hydroxymethyl)propan
Formel: C₆H₁₄O₃
Siedepunkt: 296 °C (1013 hPa)
Melting Pt: 53 °C
Dichte: 1.15 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004694
CAS-Nummer: 77-99-6
EINECS: 201-074-9

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 57 - 61 °C
Molar Mass 134.17 g/mol
Bulk Density 700 kg/m3
Boiling Point 296 - 298 °C (1013 hPa)
Vapor Pressure  - 0.1 hPa (50 °C)
Flash Point 179 °C DIN 51758
Explosion Limit 2 - 11.8 %(V)
pH 6.5 (100 g/l, H2O, 20 °C) (External MSDS)
Ignition Point 375 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR