Bis(triphenylphosphin)palladium(II)chlorid zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyms: Bis(triphenylphosphine)palladium(II) dichloride

M-Clarity™ quality level = MQ200

8.04174.0001 8.04174.0005 8.04174.8500
8.04174.0001EA 105 CHF
8.04174.0001 8.04174.0005 8.04174.8500
Bis(triphenylphosphin)palladium(II)chlorid zur Synthese, Sigma-Aldrich®
Bis(triphenylphosphin)palladium(II)chlorid
Formel: C₃₆H₃₀Cl₂P₂Pd
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009593
CAS-Nummer: 13965-03-2
EINECS: 237-744-2
UN: 1759
ADR: 8,III

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) insoluble
Molar Mass 701.91 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR