(2-Chlorpropyl)dimethylammoniumchlorid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyms: N-(2-Chloropropyl)-N,N-dimethylammonium chloride, 2-Chloro-N,N-dimethylpropan-1-amine hydrochloride, 2-Dimethylaminoisopropyl chloride hydrochloride

M-Clarity™ quality level = MQ200

8.03240.0250
8.03240.0250EA 87.1 CHF
8.03240.0250
(2-Chlorpropyl)dimethylammoniumchlorid, Sigma-Aldrich®
2-Chloropropyldimethylammonium chloride
Formel: C₅H₁₃Cl₂N
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00012534
CAS-Nummer: 4584-49-0
EINECS: 224-971-7

Order Now

Spezifikation Testergebnisse

Solubility 2000 g/l (20 °C)
Melting Point 188 - 191 °C
Molar Mass 158.07 g/mol
Bulk Density 350 kg/m3
Flash Point 220 °C
pH 5 - 6 (500 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR