N,N-Dimethylformamiddimethylacetal zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyms: 1,1-Dimethoxytrimethylamine, Esterate M, N,N-Dimethylformamide Dimethyl Acetal, 1,1-Dimethoxy-N,N-dimethylmethylamine, DMF-DMA

M-Clarity™ quality level = MQ200

8.03068.0025 8.03068.0100 8.03068.0500 8.03068.0001
8.03068.0025EA 49.3 CHF
8.03068.0025 8.03068.0100 8.03068.0500 8.03068.0001
N,N-Dimethylformamiddimethylacetal zur Synthese, Sigma-Aldrich®
N,N-Dimethylformamiddimethylacetal
Formel: C₅H₁₃NO₂
Siedepunkt: 102 °C (1013 hPa)
Dichte: 0.893 g/cm³ (20 °C)
Flash-Pt: 6 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008482
CAS-Nummer: 4637-24-5
EINECS: 225-063-3
UN: 1993
ADR: 3,II

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) (slow decomposition)
Melting Point -85 °C
Molar Mass 119.16 g/mol
Boiling Point 107 - 108 °C
Flash Point 6 °C
Refractive Index 1.3972 (20 °C, 589 nm)
Explosion Limit 1.3 - 17.7 %(V)
Density 0.90 g/cm3 (20 °C)
pH 7 (H2O)
Ignition Point 155 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR