Dimethylammoniumchlorid zS, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyms: Dimethylamine hydrochloride

MQ200

8.03052.0005 8.03052.0250 8.03052.1000 8.03052.9025
8.03052.0005EA 23.9 CHF
8.03052.0005 8.03052.0250 8.03052.1000 8.03052.9025
Dimethylammoniumchlorid zS, Sigma-Aldrich®
Dimethylammoniumchlorid
Formel: C₂H₈ClN
MW: 81.54 g/mol
Melting Pt: 170…173 °C
MDL Number: MFCD00012477
CAS-Nummer: 506-59-2
EINECS: 208-046-5
Merck Index: 13,03255

Order Now

Spezifikation Testergebnisse

Solubility 3000 g/l (20 °C)
Melting Point 160 °C
Molar Mass 81.54 g/mol
Bulk Density 500 kg/m3
Vapor Pressure  - 0.1 hPa (25 °C)
pH 5 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR