Diethylcarbonat zur Synthese, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ quality level = MQ200

8.02898.0100 8.02898.0500 8.02898.1000
8.02898.0100EA 27.8 CHF
8.02898.0100 8.02898.0500 8.02898.1000
Diethylcarbonat zur Synthese, Sigma-Aldrich®
Diethylcarbonat
Formel: C₅H₁₀O₃
Siedepunkt: 127 °C (1013 hPa)
Melting Pt: –43 °C
Dichte: 0.9751 g/cm³ (20 °C)
Flash-Pt: 33 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009107
CAS-Nummer: 105-58-8
EINECS: 203-311-1
UN: 2366
ADR: 3,III
Merck Index: 13,03815

Order Now

Spezifikation Testergebnisse

Solubility (20 °C) insoluble
Melting Point -43 °C
Molar Mass 118.13 g/mol
Boiling Point 126 °C (1013 hPa)
Vapor Pressure 11 hPa (20 °C)
Flash Point 33 °C
Refractive Index 1.384 (20 °C)
Explosion Limit 1.4 - 11.0 %(V)
Density 0.97 g/cm3 (20 °C)
pH 6.3 (1 g/l, H2O)
Ignition Point 445 °C
Evaporation number 14
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR