TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)

Lieferant: G Bioscienes

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

786-514
786-514EA 304 CHF
786-514
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)

Triton® X-100 is for solubilising membrane proteins during isolation of membrane-protein complexes.

MDL Number: MFCD00132505
CAS-Nummer: 9002-93-1
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR