Phosphatase assay kit
Lieferant: G Bioscienes
786-453
786-453EA 1210 CHF
786-453
Phosphatase assay kit
Tests Enzymtests
The Phosphatase Assay Kit is designed to measure the activity of phosphatases in biological samples and to screen for agonists and inhibitors of phosphatases. The Phosphatase Assay Kit uses para-nitrophenyl phosphate (pNPP), a chromogenic substrate for most phosphatases, including alkaline phosphatases, acid phosphatases, protein tyrosine phosphatases and serine/threonine phosphatases.
  • Colorimetric, pNPP based assay
  • Measure phosphatase activity in biological samples
  • Screen for phosphatase agonists and inhibitors
  • For the quantification of phosphatase activity
  • To screen for agonists and inhibitors of phosphatases
Order Now
TECHNISCHE DATEN
Environmentally Preferable