Toluol-α-sulfonylfluorid

Lieferant: G Bioscienes

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride

786-055
786-055EA 253 CHF
786-055
Toluol-α-sulfonylfluorid
Toluol-α-sulfonylfluorid

Inhibitor of cysteine proteases & mammalian acetylcholinesterase

Formel: C₇H₇FO₂S
Siedepunkt: 80…85 °C (0.3 mmHg)
Melting Pt: 91…92 °C
MDL Number: MFCD00007424
CAS-Nummer: 329-98-6
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR