Vanillin, Supelco®

Lieferant: Merck
Achtung

Synonyms: 4-Hydroxy-3-methoxybenzaldehyde

77634-5G
77634-5GEA 65.9 CHF
77634-5G
Vanillin, Supelco®
Vanillin
Formel: C₈H₈O₃
Siedepunkt: 285 °C (1013 hPa)
Melting Pt: ∼82 °C
Dichte: 1.056 g/cm³ (20 °C)
Flash-Pt: 153 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006942
CAS-Nummer: 121-33-5
EINECS: 204-465-2
Merck Index: 13,09998
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87b654a263df06ff19ddd79f39a3aa8aa5891fde&vid=87b654a263df06ff19ddd79f39a3aa8aa5891fde&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR