Methylenblau-Lösung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

77515-100ML-F
77515-100ML-FEA 53.3 CHF
77515-100ML-F
Methylenblau-Lösung, Sigma-Aldrich®
Methylenblau

Loefflers Methylene Blue stain is used for the identification of diphteria bacilli, since it differentiates the deeply staining metachromatic granules from the pale blue-staining cytoplasm.

Formel: C₁₆H₁₈ClN₃S
MW: 319,86 g/mol
MDL Number: MFCD00150008
CAS-Nummer: 61-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f53bcf3bb2b2419797249fc8b73cf6275cd6dbd&vid=8224fa77e250f979ae37e0948878144b7ad7a368&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR