ω-Bromacetophenon, Supelco®

Lieferant: Merck
Achtung

Synonyms: α-Bromoacetophenone, Phenacyl bromide, 2-Bromoacetophenone

M-Clarity™ quality level = MQ100

77450-10G
77450-10GEA 174 CHF
77450-10G
ω-Bromacetophenon, Supelco®
ω-Bromacetophenon
Formel: (C₆H₅)C(O)CH₂Br
MW: 199,05 g/mol
Siedepunkt: 253 °C (1013 hPa)
Melting Pt: 50 °C
Dichte: 1.647 g/cm³ (20 °C)
Flash-Pt: >112 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000195
CAS-Nummer: 70-11-1
EINECS: 200-724-9
UN: 2645
ADR: 6.1,II
Merck Index: 13,01386
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36039676c6c7d27548d3c59cf533ab49364bb9f5&vid=706078ac23e33620dfe6528a926846897a534229&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR