Supelco® TraceCERT® Organic Reference Standard, Glycine
Lieferant: Merck
76524-100MG
76524-100MGEA 103 CHF
76524-100MG
Supelco® TraceCERT® Organic Reference Standard, Glycine
Standards Organische Standards
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49981eddd4bd8b0724ef8618fa84acd401943462&vid=49981eddd4bd8b0724ef8618fa84acd401943462&prid=web&clid=SAPDAM