Gas sampling pump and tubes
Lieferant: GASTEC
GV-50PS
763-0292EA 521 CHF
763-0292
Gas sampling pump and tubes
Gas-Spürsysteme
Gas detector tube system accurately measures vapour concentration levels and slight changes in concentration level.
These detector tubes are used in experiments like respiration, photosynthesis and combustion to determine concentration level.
Bestellinformation: Includes a sampling pump, detector tubes and tips for detector tubes. More detector tubes are available separately.