n-Nonacosan, Supelco®

Lieferant: Merck

Synonyms: Nonacosane

M-Clarity™ Qualitätsstufe = MQ100

74156-1G
74156-1GEA 233 CHF
74156-1G
n-Nonacosan, Supelco®
n-Nonacosane
Formel: H₃C(CH₂)₂₇CH₃
MW: 408.8 g/mol
Siedepunkt: 440,8
Melting Pt: 63,7
Dichte: 0.8083 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009356
CAS-Nummer: 630-03-5
EINECS: 211-126-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1217d9c6cac82b0d18143935148ecb35e83f4644&vid=2d3310cad41170b11f2bdcc5eb0cf274a77f173d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR