Aufbewahrungsplatte, quadratische 1,2-ml-Wells, niedriges Profil
Lieferant: THERMO MATRIX TECHNOLOGIES
AB-1127
736-0198EA 335 CHF
736-0198
Aufbewahrungsplatte, quadratische 1,2-ml-Wells, niedriges Profil
Mikrotiterplatten Deep-Well-Mikroplatten
PP. The square well geometry of this 96-well plate makes it ideal for bacterial growth applications.
  • Low profile design maximises centrifuge capacity, allowing more plates to be loaded per run
  • U-shaped well bottom aids re-suspension
Maximum well volume (when sealed with 96-cap sealing mat): 1,0 ml
Maximum well volume (when heat sealed): 1,2 ml.
Maximum well volume (when sealed with 96-cap sealing mat): 1,0 ml