Microplates, 384-well, high content imaging film bottom, Corning®
Lieferant: Corning
4681 4518
734-2764EA 618 CHF
734-2764 CORN4518
Microplates, 384-well, high content imaging film bottom, Corning®
Mikrotiterplatten HTS-Mikrotiterplatten
With an ultraclear film bottom and excellent flatness (whole plate and intra-well), these microplates are ideal for high resolution cellular imaging applications.
  • COC material allows for broad chemical resistance (including DMSO) and high mechanical stability
  • Ultra-clear film with 127 µm thickness is well suited for imaging micropscopy
  • Inter- and intra-well film bottom flatness within 50 and 10 µm, respectively, optimised for high content applications
  • Low auto-fluorescence and birefringence
The microplate and film are manufactured from cyclic olefinMore Product Information