Individually Wrapped Serological Pipettes, Falcon®
Lieferant: Corning
356525 356543 356551 357507 357521 357525 357535 357543 357550 357551 357600 356507 356521 356535 356550
734-0307EA 237 CHF
734-0307 734-0313 734-0315 734-0336 734-0340 734-0343 734-0347 734-0350 734-0351 734-0352 734-0355 734-0297 734-0305 734-0310 734-0314
Individually Wrapped Serological Pipettes, Falcon®
Pipetten Serologische Pipetten
The pipette graduation alignment system is a fast and simple way to bring graduations to your line of sight. Polystyrene, disposable pipettes for tissue culture, bacteriological, and research applications.
  • Sterile
  • RNase-/DNase-free
  • Noncytotoxic, nonpyrogenicity tested to less than 0,1 EU/ml
  • Color-coded package and markings for ease of identification
Simply pop open or peel back the top of the pipette package and line up the pipettor before completely removing the pipette from its wrapping. When you remove the pipette from its package, the graduationsMore Product Information