Sodium ionophore I, Selectophore™, Funktion getestet Ionophor, Supelco®

Lieferant: Merck

Synonyms: N,N′,N′′-Triheptyl-N,N′,N′′-trimethyl-4,4′,4′′-propylidynetris(3-oxabutyramide), ETH 227

M-Clarity™ quality level = MQ100

71732-50MG
71732-50MGEA 3240 CHF
71732-50MG
Sodium ionophore I, Selectophore™, Funktion getestet Ionophor, Supelco®
Sodium ionophore I

Neutral ionophore for liquid-membrane electrodes of high selectivity for Na+


Verpackung: Clear borosilicate glass, high hydrolytic resistance (Type I)

Formel: C₃₆H₇₁N₃O₆
MDL Number: MFCD00042950
CAS-Nummer: 61183-76-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04855990850be9c792e9d95892adca7193da2a36&vid=7e67c89101c11696e23c1df228da5d88e90e039c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR