Automatic colony counter, aCOLyte 3 HD
Lieferant: SYNBIOSIS
ACOL3-HD
710-1970EA 8060 CHF
710-1970
Automatic colony counter, aCOLyte 3 HD
Kolonienzähler
aCOLyte 3 HD uses a powerful algorithm for colony counting on pour plates and Spiral® plates.
  • Automated in one configuration, uses powerful algorithm
  • Sensitive CCD camera and easy-to-use camera
  • Strong light eliminated by a detachable screen
The plates are automatically counted with results transferred to Excel or Open Office. This can be used with any conforming laptop or desktop PC and connects by using a USB port.