Dichlormethan, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyms: Methylene chloride

7053.2500EA 133 CHF
7053.2500 7053.1000 7053.9025 7053.9200
Dichlormethan, BAKER ANALYZED® ACS, J.T.Baker®
Dichlormethan
Formel: CH₂Cl₂
MW: 84.93 g/mol
Siedepunkt: 39.8
Melting Pt: –95 °C
Dichte: 1.322 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000881
CAS-Nummer: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Exceeds ACS Specifications
Assay (by GC) (corrected for water) min. 99.5 %
Appearance clear
Color (APHA) max. 10
Free Halogens passes test
Residue after Evaporation max. 0.002 %
Titrable Acid (meq/g) max. 0.0002
Water (H2O) max. 0.02 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR