Milchsäure ≥99%, nach Ph. Eur. getestet, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

69775-1L 69775-250ML
69775-1LEA 188.87 CHF
69775-1L 69775-250ML
Milchsäure ≥99%, nach Ph. Eur. getestet, Fluka™
Milchsäure
Formel: H₃CCH(OH)COOH
Siedepunkt: 122 °C (20 hPa)
Dichte: 1.209 g/cm³
Flash-Pt: 113 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004520
CAS-Nummer: 50-21-5
Merck Index: 13,05351

Order Now

Spezifikation Testergebnisse

Appearance of the substance complying
Assay of total acid 88.0 - 92.0 %
Sulfated ash Max. 0.1 %
Calcium (Ca) Max. 200 ppm
Heavy metals (as Pb) Max. 10 ppm
Sulfate (SO₄) Max. 200 ppm
Citronellic-, Oxalic- and Phosphoric acid complying
Sugar and other reducing substances complying
In ether insoluble matter complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR