Uracil, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyms: Pyrimidine-2,4(1H,3H)-dione

6630-10
6630-10EA 102 CHF
6630-10
Uracil, Millipore®
Uracil
Formel: C₄H₄N₂O₂
MW: 112,09 g/mol
Siedepunkt: 379 °C (1013 hPa)
Melting Pt: 194 °C
Dichte: 0.99744 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006016
CAS-Nummer: 66-22-8
EINECS: 200-621-9
Merck Index: 12,09985

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR