Chloroform ≥99,9% stabilisiert, CHROMASOLV™ Plus für die HPLC, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Stabilised with ethanol 0.5-1.0%

650471-1L
650471-1LEA 177.13 CHF
650471-1L
Chloroform ≥99,9% stabilisiert, CHROMASOLV™ Plus für die HPLC, Riedel-de Haën™
Chloroform

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities


Beilstein Number: 1731042

Formel: CHCl₃
Siedepunkt: 60-62 °C (1.013 hPa)
Melting Pt: -63 °C
Dichte: 1.48 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000826
CAS-Nummer: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Identity (IR) complying
Purity excluding stabilizer (GC) Min. 99.90 %
Ethanol (GC) 0.5 - 1.0 %
Non-volatile matter Max. 0.0001 %
Water (Karl Fischer) Max. 0.02 %
Absorbance at 400 nm Max. 0.005
Absorbance at 275 nm Max. 0.005
Absorbance at 250 nm Max. 0.30
Absorbance at 245 nm Max. 1.00
GC (as heptachlor epoxide) Max. 10 ng/l
Fluorescence (chinin) at 365 nm Max. 0.5 ppb
Contains 0.5-1.0% ethanol as stabilizer
Vapor Density 4.1 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR