Digital refractometers
Lieferant: A.KRÜSS Optronic
DR6000 DR6000-T DR6100 DR6100-T DR6100-TF DR6200 DR6200-T DR6200-TF DR6300 DR6300-T DR6300-TF
635-0087EA 0 CHF
635-0087 635-0089 635-0091 635-0093 635-0094 635-0095 635-0097 635-0098 635-0099 635-0101 635-0102
Digital refractometers
Refraktometer Tischrefraktometer
These digital refractometers are fitted with integrated Peltier thermostats for outstanding temperature control. Measurements are made with high accuracy and are not affected by colour or cloudiness.
  • Touch screen with clear menu navigation
  • High quality ceramic PT100 sensor
  • Full GLP suitability with user management functionality
  • Data export in Excel format to a USB flash drive
  • Large measurement range with high resolution
Built-in electronic Peltier thermostat (DR6200-T or DR6300-TF) with a sample temperature control range 10 to 80 °C (stability is 0,05More Product Information