Upright Microscopes, Biological, Panthera E2, Motic
Lieferant: MOTIC
1100104601112 1100104601122 1100104601111 1100104601121
630-3448EA 2030 CHF
630-3448 630-3450 630-3447 630-3449
Upright Microscopes, Biological, Panthera E2, Motic
Mikroskope
Panthera E2 microscope combine high quality materials and precise engineering, user-friendly design. With a 360º rotatable head specially for space-saving storage, and an extreme efficient low power illumination, allowing it to run on a Mobile Battery Pack for several hours. It has been designed for Academic Lab demand.
  • 360º rotatable head (storage saving space)
  • Plan Achromatic UC objectives, (good optic quality)
  • 2 Integrated USB ports: 1 for camera power and 1 for external mobile battery
  • Fixed khoeler LED ilumination
  • Can be powered by regular cable 110 - 2
More Product Information